Phalloidin, XFD488
Lieferant: AAT BIOQUEST
23153
AATB23153EA 460 EUR
AATB23153
Phalloidin, XFD488
Proteine und Peptide
XFD488 has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
  • Well-cited conjugate
  • High affinity and selectivity
  • In house quality control
Bitte beachten For research use only.