Phalloidin, XFD594
Lieferant: AAT BIOQUEST
23158
AATB23158EA
460
EUR
AATB23158
Phalloidin, XFD594
Proteine und Peptide
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Well-cited conjugate
- High affinity and selectivity
- In house quality control
Bitte beachten For research use only.