Texas Red®-DHPE ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
(N-(Texas Red® sulphonyl)-1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt
23300
AATB23300EA
152
EUR
AATB23300
Texas Red®-DHPE ≥95%
Texas Red®-DHPE
Texas Red® DHPE is a phospholipid that is labeled on the head group with the bright, red-fluorescent Texas Red® dye (excitation/emission maxima ~595/615 nm).
Formel:
C₇₄H₁₁₇N₄O₁₄PS₂ Molecular Weight: 1380,77 g/mol |
CAS-Nummer:
187099-99-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...