TRITC-DHPE ≥90%

Lieferant: AAT BIOQUEST

Synonyme: N-(tetramethylrhodamine-6-thiocarbamoyl)-1,2-dihexadecanoyl- sn-glycero-3-phosphoethanolamine, triethylammonium salt

23301
AATB23301EA 152 EUR
AATB23301
TRITC-DHPE ≥90%
TRITC-DHPE

TRITC DHPE is a phospholipid that is labeled on the head group with the bright, red-fluorescent TRITC dye.


  • Cell Tracer
  • Long Emission Wavelength


Bitte beachten For research use only.

Formel: C₆₈H₁₁₀N₅O₁₁PS
Molecular Weight: 1236,67 g/mol
CAS-Nummer: 176181-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR