OG488 DHPE
Lieferant: AAT BIOQUEST
23302
AATB23302EA 136 EUR
AATB23302
OG488 DHPE
OG488 DHPE
OG488 DHPE is a phospholipid that is labeled on the head group with the bright, green-fluorescent fluorinated fluorescein dye (also called Oregon Green® 488, Oregon Green® is a trademark of Invitrogen).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only