Fluorescein-DHPE ≥90%
Lieferant: AAT BIOQUEST
Synonyme:
Fluorescein DHPE, N-(fluorescein-5-thiocarbamoyl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt
23304
AATB23304EA
152
EUR
AATB23304
Fluorescein-DHPE ≥90%
Fluorescein-DHPE
Fluorescein DHPE is a phospholipid that is labeled on the head group with the bright, green-fluorescent fluorescein dye.
- Bright Fluorescence
- Robust
Bitte beachten For research use only.
Formel:
C₆₄H₁₀₀N₃O₁₃PS Molecular Weight: 1182,53 g/mol |
CAS-Nummer:
87706-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...