Tide Fluor™ 7WS amine, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF7WS amine

2331
AATB2331EA 307 EUR
AATB2331
Tide Fluor™ 7WS amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 7WS amine

Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1047,07 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94233e614b6cedb9457fc0764b418817c14df6e6&vid=b5db3de5e043edc354e1a37c9d9a7cd67e937bf2&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR