Tide Fluor™ 7WS, succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF7WS SE
2333
AATB2333EA
152
EUR
AATB2333
Tide Fluor™ 7WS, succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 7WS, succinimidyl ester
Tide Fluor™ 7WS (TF7WS) family has the spectral properties similar to those of Cy7, IRDye 800 and Alexa Fluor 750.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
1076,39 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...