Tide Fluor™ 8WS acid, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF8WS acid

2335
AATB2335EA 307 EUR
AATB2335
Tide Fluor™ 8WS acid, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 8WS acid

Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1057,11 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91b8e883414927c709ebf7f3add12856d48b5c1a&vid=50d050d3870e74b58fb1cdc40ab6cee604aef110&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR