Tide Fluor™ 8WS amine, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF8WS amine

2336
AATB2336EA 307 EUR
AATB2336
Tide Fluor™ 8WS amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 8WS amine

Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1213.22 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e990ca1bbe1f5ff89ffe713bf79c18cf35af5178&vid=506d43357d0f4ed92a5465cb05eafbb8fced09e7&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR