Tide Fluor™ 8WS maleimide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF8WS maleimide

2337
AATB2337EA 307 EUR
AATB2337
Tide Fluor™ 8WS maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 8WS maleimide

Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1439,86 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2bfc84459255e61fa223b0d673f271703c18952&vid=3bcbaa81704ad6a3f9d8f80eea060840c995b3fa&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR