Tide Fluor™ 8WS, succinimidyl ester, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF8WS SE

2338
AATB2338EA 152 EUR
AATB2338
Tide Fluor™ 8WS, succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 8WS, succinimidyl ester

Tide Fluor™ 8WS (TF8WS) family has the spectral properties similar to those of IRDye 800.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1343,73 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aa6b7cf63e6405ec696a7fd1a34f0378882161a&vid=a6fa43a0270e386efa8f13196388c5da1899f90c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR