Tide Fluor™ 3WS maleimide, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF3WS maleimide
2344
AATB2344EA
307
EUR
AATB2344
Tide Fluor™ 3WS maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3WS maleimide
Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
961 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...