Tide Fluor™ 3WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF3WS SE

2346
AATB2346EA 460 EUR
AATB2346
Tide Fluor™ 3WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 3WS succinimidyl ester

Tide Fluor™ 3WS (TF3WS) family has the spectral properties essentially identical to those of Cy3.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 1024,31 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2a5f9306a8c36265e010828e511e030346dd5fb&vid=2a0d1a0a69974e135db3de2a9ca116af44ec2aac&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR