Tide Fluor™ 2WS acid, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF2WS acid
2348
AATB2348EA
614
EUR
AATB2348
Tide Fluor™ 2WS acid, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2WS acid
Tide Fluor™ 2WS (TF2WS) family has the spectral properties similar to those of fluorescein, DyLight 488 and Alexa Fluor 488.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
1113,18 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...