Tide Fluor™ 2WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF2WS SE
2349
AATB2349EA
614
EUR
AATB2349
Tide Fluor™ 2WS succinimidyl ester, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2WS succinimidyl ester
Tide Fluor™ 2WS (TF2WS) family has the spectral properties similar to those of fluorescein, DyLight 488 and Alexa Fluor 488.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
945,07 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...