5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) Gemisch aus Isomeren ≥95%

Lieferant: AAT BIOQUEST

235
AATB235EA 152 EUR
AATB235
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) Gemisch aus Isomeren ≥95%
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF)

CDCF [5(6)-carboxy-2',7'-dichlorofluorescein] is the fluorescent product of 5(6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate and 5(6)-carboxy-2',7'-dichlorofluorescein diacetate in cells.


  • ROS Indicator
  • Stable Fluorescence


Bitte beachten For research use only.

Formel: C₂₁H₁₀Cl₂O₇
MDL: MFCD00037452
CAS-Nummer: 111843-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR