Tide Fluor™ 2WS Amine, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF2WS amine

2351
AATB2351EA 307 EUR
AATB2351
Tide Fluor™ 2WS Amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 2WS Amine

Tide Fluor™ 2WS (TF2WS) family has the spectral properties similar to those of fluorescein, DyLight 488 and Alexa Fluor 488.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 805,61 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db9ba81e7e30dbe678728adf05232a422d38215e&vid=4b889a498bb8d13651eb3375df5c47651d63a4f8&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR