6-OG488 azide
Lieferant: AAT BIOQUEST
236
AATB236EA 206 EUR
AATB236
6-OG488 azide
6-OG488 azide
6-OG488 azide has the same chemical structure to that of Oregon Green™ 488 azide.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only