AzoDye-1 carboxylic acid
Lieferant: AAT BIOQUEST
2400
AATB2400EA
442
EUR
AATB2400
AzoDye-1 carboxylic acid
AzoDye-1 Carboxylic acid
AzoDye-1 Carboxylic Acid is an excellent building block for labeling amino-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only