MycoLight™ Green JJ98 *5 mM in DMSO*
Lieferant: AAT BIOQUEST
24000
AATB24000EA
307
EUR
AATB24000
MycoLight™ Green JJ98 *5 mM in DMSO*
Tests
Zelltests
MycoLight™ Green JJ98 is a green-fluorescent nuclear and chromosome stain that is permeant to both prokaryotic and eukaryotic cell membranes.
- Cell-Permeable
- High DNA Affinity
- Bright Fluorescence
Bitte beachten For research use only.