MycoLight™ Green JJ99 *5 mM in DMSO*
Lieferant: AAT BIOQUEST
24001
AATB24001EA 307 EUR
AATB24001
MycoLight™ Green JJ99 *5 mM in DMSO*
Tests Zelltests
MycoLight™ Green JJ99 stain is an excellent green-fluorescent nuclear and chromosome counterstain that is permeant to both prokaryotic and eukaryotic cell membranes.
  • Cell-Permeable
  • High DNA Affinity
  • Bright Fluorescence
Bitte beachten For research use only.