AzoDye-1 succinimidyl ester
Lieferant: AAT BIOQUEST
2402
AATB2402EA
144
EUR
AATB2402
AzoDye-1 succinimidyl ester
AzoDye-1 Succinimidyl ester
AzoDye-1 Succinimidyl Ester is an excellent building block for labeling amino-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only