AzoDye-1 maleimide
Lieferant: AAT BIOQUEST
2404
AATB2404EA
144
EUR
AATB2404
AzoDye-1 maleimide
AzoDye-1 Maleimide
AzoDye-1 maleimide is an excellent building block for labeling thiol-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only