AzoDye-1 CPG 500 A (500 A)
Lieferant: AAT BIOQUEST
2408
AATB2408EA 295 EUR
AATB2408
AzoDye-1 CPG 500 A (500 A)
AzoDye-1 CPG
AzoDye-1 CPG is an excellent building block for preparing FRET oligo molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only