AzoDye-1 CPG 1000 A (1000 A)
Lieferant: AAT BIOQUEST
2410
AATB2410EA 295 EUR
AATB2410
AzoDye-1 CPG 1000 A (1000 A)
AzoDye-1 CPG
AzoDye-1 CPG is an excellent building block for preparing FRET oligo molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only