AzoDye-1 Peg10 alkyne
Lieferant: AAT BIOQUEST
2415
AATB2415EA
212
EUR
AATB2415
AzoDye-1 Peg10 alkyne
AzoDye-1 PEG10-Alkyne
AzoDye-1 PEG10 alkyne is an excellent building block for labeling azide-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only