AzoDye-2 carboxylic acid
Lieferant: AAT BIOQUEST
2420
AATB2420EA
442
EUR
AATB2420
AzoDye-2 carboxylic acid
AzoDye-2 Carboxylic acid
AzoDye-2 Carboxylic Acid is an excellent building block for labeling amino-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only