MycoLight™ PMA [Propidium Monoazide] 5 mM
Lieferant: AAT BIOQUEST
24216
AATB24216EA
198
EUR
AATB24216
MycoLight™ PMA [Propidium Monoazide] 5 mM
MycoLight™ PMA [Propidium Monoazide]
Propidium monoazide (PMA) is one of the most common vPCR reagents for the selective detection of DNA of live cells.
- Stable for 24 months
The common PCR assays have been routinely used for simple, sensitive, and specific pathogen identification.
Bitte beachten For Research Use Only