AzoDye-2 succinimidyl ester
Lieferant: AAT BIOQUEST
2422
AATB2422EA 144 EUR
AATB2422
AzoDye-2 succinimidyl ester
AzoDye-2 Succinimidyl ester
AzoDye-2 Succinimidyl Ester is an excellent building block for labeling amino-containing molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only