AzoDye-2 maleimide
Lieferant: AAT Bioquest
2424
AATB2424EA 144 EUR
AATB2424
AzoDye-2 maleimide
AzoDye-2 Maleimide
AzoDye-2 maleimide is an excellent building block for labeling thiol-containing molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only