AzoDye-2 amine
Lieferant: AAT BIOQUEST
2426
AATB2426EA
144
EUR
AATB2426
AzoDye-2 amine
AzoDye-2 Amine
AzoDye-2 amine is an excellent building block for labeling carbonyl-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only