AzoDye-2 CPG 500 A (1000 A)
Lieferant: AAT BIOQUEST
2430
AATB2430EA
295
EUR
AATB2430
AzoDye-2 CPG 500 A (1000 A)
AzoDye-2 CPG
AzoDye-2 CPG is an excellent building block for preparing FRET oligo molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only