AzoDye-2 C2 azide
Lieferant: AAT BIOQUEST
2431
AATB2431EA 136 EUR
AATB2431
AzoDye-2 C2 azide
AzoDye-2 C2 Azide
AzoDye-2 C2 azide is an excellent building block for labeling alkyne-containing molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten Research use only (RUO).