AzoDye-2 alkyne
Lieferant: AAT Bioquest
2434
AATB2434EA 144 EUR
AATB2434
AzoDye-2 alkyne
AzoDye-2 Alkyne
AzoDye-2 alkyne is an excellent building block for labeling azide-containing molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only