AzoDye-2 PEG10 alkyne
Lieferant: AAT BIOQUEST
2435
AATB2435EA
212
EUR
AATB2435
AzoDye-2 PEG10 alkyne
AzoDye-2 PEG10 alkyne
AzoDye-2 PEG10 alkyne is an excellent building block for labeling azide-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only