Trypan sodium salt 0.1 M, Trypan UltraBlue™ zur Markierung toter Zellen

Lieferant: AAT BIOQUEST
Achtung

2455
AATB2455EA 307 EUR
AATB2455
Trypan sodium salt 0.1 M, Trypan UltraBlue™ zur Markierung toter Zellen
Trypan sodium salt

Trypan UltraBlue™ is similar to trypan blue in cell permeability.


  • Minimal cytotoxicity
  • High purity

Molecular Weight: 1000 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR