Phenol Red, sodium salt

Lieferant: AAT Bioquest
Achtung

2460
AATB2460EA 307 EUR
AATB2460
Phenol Red, sodium salt
Phenol Red, sodium salt

Phenol red (also known as phenolsulfonphthalein or PSP) is a pH indicator that is frequently used in cell biology laboratories.

Formel: C₁₉H₁₃O₅SNa
CAS-Nummer: 34487-61-6
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR