AzoDye-3 maleimide
Lieferant: AAT Bioquest
2474
AATB2474EA 136 EUR
AATB2474
AzoDye-3 maleimide
AzoDye-3 Maleimide
AzoDye-3 maleimide is an excellent building block for labeling thiol-containing molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only