AzoDye-3 CPG 500 A (1000 A)
Lieferant: AAT BIOQUEST
2480
AATB2480EA 275 EUR
AATB2480
AzoDye-3 CPG 500 A (1000 A)
AzoDye-3 CPG
AzoDye-3 CPG is an excellent building block for preparing FRET oligo molecules.
  • Stable for 12 months
Quality tested in-house
Bitte beachten Research use only (RUO).