AzoDye-3 azide
Lieferant: AAT BIOQUEST
2482
AATB2482EA
136
EUR
AATB2482
AzoDye-3 azide
AzoDye-3 Azide
AzoDye-3 azide is an excellent building block for labeling alkyne-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only