AzoDye-3 alkyne
Lieferant: AAT BIOQUEST
2484
AATB2484EA
136
EUR
AATB2484
AzoDye-3 alkyne
AzoDye-3 Alkyne
AzoDye-3 alkyne is an excellent building block for labeling azide-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten Research use only (RUO).