AzoDye-3 PEG10 alkyne
Lieferant: AAT BIOQUEST
2485
AATB2485EA
206
EUR
AATB2485
AzoDye-3 PEG10 alkyne
AzoDye-3 PEG10 Alkyne
AzoDye-3 PEG10 alkyne is an excellent building block for labeling azide-containing molecules.
- Stable for 12 months
Quality tested in-house
Bitte beachten Research use only (RUO).