mFluor™ Violet 500-Wheat Germ Agglutinin (WGA) Conjugate
Lieferant: AAT BIOQUEST
25452
AATB25452EA
202
EUR
AATB25452
mFluor™ Violet 500-Wheat Germ Agglutinin (WGA) Conjugate
Proteinkonjugate und Markierungen
mFluor™ Violet 500 labelled WGA is well-excited by the violet laser, emitting a bright green fluorescence at 501 nm.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Bitte beachten For research use only.