mFluor™ Violet 540-Wheat Germ Agglutinin (WGA) Conjugate
Lieferant: AAT BIOQUEST
25454
AATB25454EA 202 EUR
AATB25454
mFluor™ Violet 540-Wheat Germ Agglutinin (WGA) Conjugate
Proteinkonjugate und Markierungen
mFluor™ Violet 540 labelled WGA is well-excited by the violet laser, emitting a bright green fluorescence at 535 nm.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Bitte beachten For research use only.