Cyanine 5 potassium salt ≥95% [equivalent to Cy5® potassium salt]
Lieferant: AAT BIOQUEST
255
AATB255EA 307 EUR
AATB255
Cyanine 5 potassium salt ≥95% [equivalent to Cy5® potassium salt]
Cyanine 5 potassium salt
A variety of cyanine 5 (Cy5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
  • Multiple Bioconjugation Applications
  • Excellent Quality
Bitte beachten For research use only.
CAS-Nummer: 449175-58-0