Wheat Germ Agglutinin, XFD532 Labelled *XFD532 Same Structure to Alexa Fluor® 532*
Lieferant: AAT BIOQUEST
25502
AATB25502EA
144
EUR
AATB25502
Wheat Germ Agglutinin, XFD532 Labelled *XFD532 Same Structure to Alexa Fluor® 532*
Proteinkonjugate und Markierungen
XFD532, manufactured by AAT Bioquest, has an identical chemical structure to Alexa Fluor® 532, a trademark of ThermoFisher.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.