Wheat Germ Agglutinin, XFD594
Lieferant: AAT BIOQUEST
25509
AATB25509EA 143 EUR
AATB25509
Wheat Germ Agglutinin, XFD594
Proteine und Peptide
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
  • Well-cited conjugate
  • High affinity and selectivity
  • Multiple applications
Bitte beachten For research use only.