Wheat Germ Agglutinin, XFD647 Labelled *XFD647 Same Structure to Alexa Fluor® 647*
Lieferant: AAT BIOQUEST
25512
AATB25512EA
144
EUR
AATB25512
Wheat Germ Agglutinin, XFD647 Labelled *XFD647 Same Structure to Alexa Fluor® 647*
Proteinkonjugate und Markierungen
XFD647, manufactured by AAT Bioquest, has an identical chemical structure to Alexa Fluor® 647, a trademark of ThermoFisher.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.