Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 647
Lieferant: AAT BIOQUEST
25559
AATB25559EA
143
EUR
AATB25559
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 647
Proteine und Peptide
Wheat germ agglutinin (WGA) is a lectin that binds to N-acetyl-D-glucosamine and sialic acid.
- Glycoconjugate binding
- Very bright signal
- Multiple applications
Bitte beachten For research use only.